Order Entry
Switzerland
ContactUsLinkComponent
 
 
Magnesium nitrate hexahydrate, Suprapur® for trace analysis, Supelco®
 :  Merck
 
 
Magnesium nitrate hexahydrate, Suprapur® for trace analysis, Supelco®
 :  Merck
 

 

  • Solubility:
    420 g/l (20 °C)
  • Melting Point:
    89 °C (for the hexahydrate)
  • Molar Mass:
    256.41 g/mol
  • Density:
    1.46 g/cm3 (20 °C)
  • pH:
    5.0 - 7.0 (50 g/l, H2O, 25 °C)

 

Frequently Bought Together