Order Entry
Switzerland
ContactUsLinkComponent
Magnesium chloride hexahydrate, crystallised, EMPROVE® EXPERT Ph. Eur., BP, USP, JPC, ACS, SAFC®
Magnesium chloride hexahydrate, crystallised, EMPROVE® EXPERT Ph. Eur., BP, USP, JPC, ACS, SAFC®
Catalog # 1.37008.6025
CAS Number:  
Magnesium chloride hexahydrate, crystallised, EMPROVE® EXPERT Ph. Eur., BP, USP, JPC, ACS, SAFC®
Catalog # 1.37008.6025
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay (complexometric):
    99.0 - 101.0 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • Insoluble matter:
    ≤ 0.005 %
  • Acidity or alkalinity:
    passes test
  • pH-value (5 %; water):
    5.0 - 7.0
  • Bromide (Br):
    ≤ 0.05 %
  • Nitrate (NO3) (**):
    ≤ 0.001 %
  • Phosphate (PO4) (**):
    ≤ 0.0005 %
  • Sulfate (SO₄):
    ≤ 0.002 %
  • Heavy metals (as Pb):
    ≤ 0.0005 %
  • Al (Aluminium):
    ≤ 0.0001 %
  • As (Arsenic):
    ≤ 0.0002 %
  • Ba (Barium):
    passes test
  • Ba (Barium) (**):
    ≤ 0.005 %
  • Ca (Calcium):
    ≤ 0.01 %
  • Fe (Iron):
    ≤ 0.0005 %
  • Hg (Mercury):
    ≤ 0.0001 %
  • K (Potassium):
    ≤ 0.0050 %
  • Mn (Manganese) (**):
    ≤ 0.0005 %
  • Na (Sodium) (**):
    ≤ 0.005 %
  • NH₄ (Ammonium):
    ≤ 0.002 %
  • Pb (Lead):
    ≤ 0.0004 %
  • Sr (Strontium) (**):
    ≤ 0.005 %
  • Residual solvents (Ph Eur/USP/ICH):
    excluded by manufacturing process
  • Water:
    51.0 - 55.0 %
  • Endotoxins:
    ≤ 3.0 I.U./g
  • Microbiological test (Total aerobic microbial count (TAMC)):
    ≤ 100 CFU/g
  • Microbiological test (Total yeast and mould count (TYMC)):
    ≤ 100 CFU/g

Specification Test Results

About this item

 :
 :203.3 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:117 °C
Density:1.57 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7791-18-6
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781