Order Entry
Switzerland
ContactUsLinkComponent
Calcium chloride dihydrate, granular USP, Multi-Compendial, J.T.Baker®
Irritante
Calcium chloride dihydrate, granular USP, Multi-Compendial, J.T.Baker®
Codice catalogo 1335-07
Nome del fornitore:  Avantor
Numero CAS:  
undefined
Irritante
Calcium chloride dihydrate, granular USP, Multi-Compendial, J.T.Baker®
Codice catalogo 1335-07
Nome del fornitore:  Avantor
Numero CAS:  

Specifiche

  • Pack type:
    Poly pail, screw top
  • Pk:
    12 kg
  • Seq:
    4187

Specifiche

Risultati dei test sulle specifiche

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Identification:
    Passes Test
  • USP - pH of 5% Solution at 25°C:
    4.5 - 9.2
  • USP - Iron, Aluminum, and Phosphate:
    Passes Test
  • USP - Magnesium and Alkali Salts:
    ≤ 1.0 %
  • USP - Assay (CaCl₂·2H₂O):
    99.0 - 107.0 %
  • Endotoxin Concentration (EU/g):
    ≤ 10
  • EP/BP - Assay (CaCl₂·2H₂O):
    97.0 - 103.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Acidity or Alkalinity:
    Passes Test
  • EP/BP - Sulfate (SO₄):
    ≤ 300 ppm
  • EP/BP - Aluminum (Al):
    Passes Test
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Magnesium and Alkali Metals:
    ≤ 0.5 %
  • JP - Assay (CaCl₂·2H₂O):
    96.7 - 103.3 %
  • JP - Identification:
    Passes Test
  • JP - pH (1 in 20):
    4.5 - 9.2
  • JP - Heavy Metals (as Pb):
    ≤ 10 ppm
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Sulfate (SO₄):
    ≤ 0.024 %
  • JP - Hypochlorite:
    Passes Test
  • JP - Iron, Aluminum or Phosphate:
    Passes Test
  • JP - Barium (Ba):
    Passes Test
  • JP - Arsenic (As):
    ≤ 2 ppm
  • No Class 1,2,3 or other solvents are used or produced in the:
    Passes Test
  • manufacturing or purification of the product.:
    = 2 ppm

Risultati dei test sulle specifiche

Informazioni su questo articolo

 :
 :147.01 g/mol
IndexMerck:13,01660
Punto di ebollizione:1600 °C (1013 hPa)
Punto di fusione:176 °C
Densità:1.85 g/cm³ (20 °C)
 :Management of Change (MOC) category = R
temperatura di conservazione:Ambient
EINECS:233-140-8
 :GHS07
Numero CAS:10035-04-8
Formula:CaCl₂·2H₂O
Numero MDL:MFCD00149613