Order Entry
Switzerland
ContactUsLinkComponent
 
Aluminium potassium sulphate dodecahydrate 99.5-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Catalog # 21110.296
Supplier:  VWR Chemicals
CAS Number:  
undefined
Aluminium potassium sulphate dodecahydrate 99.5-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Catalog # 21110.296
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    99.5 - 100.5 %
  • Appearance of solution:
    Passes test Ph.Eur.
  • Identification A:
    Passes test Ph.Eur.
  • Identification B:
    Passes test Ph.Eur.
  • Identification C:
    Passes test Ph.Eur.
  • pH (20°C; 10 %):
    3.0 - 3.5
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • NH4 (Ammonium):
    Max. 50 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Na (Sodium):
    Max. 0.02 %
  • Pb (Lead):
    Max. 5 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test

Specification Test Results

About this item

 :
 :474.39 g/mol
Merck Index:12,00373
Melting Pt:92.5 °C
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-141-3
REACH:01-2119960162-44
CAS Number:7784-24-9
Formula:AlK(SO₄)₂·12H₂O
MDL Number:MFCD00149143