
Lithium sulphate monohydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10102-25-7
:
Formula:Li₂SO₄·H₂O
MDL Number:MFCD00149766
:127.96 g/mol
Merck Index:14,5541
Melting Pt:130 °C (-H₂O)
Density:2.06
EINECS:233-820-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together