Order Entry
Switzerland
ContactUsLinkComponent
Sodium molybdate(VI) dihydrate ≥99.5% ACS
Sodium molybdate(VI) dihydrate ≥99.5% ACS
Catalog # 0541-250G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium molybdate(VI) dihydrate ≥99.5% ACS
Catalog # 0541-250G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Ammonium:
    <0.001%
  • Chloride:
    ≤0.005%
  • Heavy Metals (as Pb):
    ≤0.0005%
  • Insolubles:
    ≤0.005%
  • Iron:
    ≤0.001%
  • pH (5%, Water) @25 °C:
    7 – 10.5
  • Phosphate:
    ≤0.0005%
  • Purity:
    99.5 – 103%
  • Sulphate:
    ≤0.015%

Specification Test Results

About this item

 :
 :241.95 g/mol
Merck Index:13,08718
Melting Pt:687 °C
Density:3.28 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-551-7
CAS Number:10102-40-6
Formula:Na₂MoO₄·2H₂O
MDL Number:MFCD00149170