Order Entry
Switzerland
ContactUsLinkComponent
Potassium hexacyanoferrate(II) trihydrate ≥96%, TECHNICAL
Environment
Potassium hexacyanoferrate(II) trihydrate ≥96%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Environment
Potassium hexacyanoferrate(II) trihydrate ≥96%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 96 %

Specification Test Results

Frequently Bought Together