Order Entry
Switzerland
ContactUsLinkComponent
5-Sulphosalicylic acid dihydrate ≥99%, AnalaR NORMAPUR® analytical reagent
Irritant
5-Sulphosalicylic acid dihydrate ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
5-Sulphosalicylic acid dihydrate ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Free salicylic acid:
    Max. 0.04 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 0.1 %
  • Insolubility in NH3 4 N:
    Max. 0.03 %
  • Insolubility in ethanol 96 % vol:
    Max. 0.1 %
  • Insolubility in water:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • Fe (Iron):
    Max. 25 ppm

Specification Test Results

Frequently Bought Together