tri-Sodium citrate dihydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, E331, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:6132-04-3
:
Formula:Na₃C₆H₅O₇·2H₂O
MDL Number:MFCD00150031
:294.1 g/mol
Merck Index:13,08675
Melting Pt:150 °C
Density:1.76 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-675-3
Related Information
Certificates
Search by Lot NumberMERP1.06447.5000_TDS_EN ProductPage_106447
Product specification
msds_106447
SDS
Frequently Bought Together