tri-Sodium citrate dihydrate 99.0-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Katalognr. 3648-07
Anbieter: Avantor
CAS-Nummer:
Technische Daten
- Pack type:PE drum, open head
- Pk:12 kg
- Seq:4187
Technische Daten
Ergebnisse des Spezifikationstests
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- USP - Alkalinity:Passes Test
- USP - Water (H₂O):10.0 - 13.0 %
- USP - Tartrate:Passes Test
- USP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):99.0 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Readily Carbonizable Substances:Passes Test
- EP/BP - Chloride (Cl):≤50 ppm
- EP/BP - Oxalate (C₂O₄):≤300 ppm
- EP/BP - Sulfate (SO₄):≤150 ppm
- EP/BP - Water (H₂O):11.0 - 13.0 %
- EP/BP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):99.0 - 101.0 %
- EP/BP - Endotoxin Concentration (EU/g):≤2.5
- JP - Assay (C₆H₅Na₃O₇) (calculated on the anhydrous basis):99.0 - 101.0 %
- JP - pH (1 in 20):7.5 - 8.5
- JP - Clarity and Color of Solution:Passes Test
- JP - Chloride (Cl):≤0.015 %
- JP - Identification:Passes Test
- JP - Sulfate (SO₄):≤0.048 %
- JP - Heavy Metals (as Pb):≤10 ppm
- JP - Arsenic (As):≤2 ppm
- JP - Tartrate:Passes Test
- JP - Oxalate (C₂O₄):Passes Test
- JP - Readily Carbonizable Substances:Passes Test
- JP - Loss on Drying at 180°C:10.0 - 13.0 %
- Calcium (Ca):≤0.005 %
- Iron (Fe):≤5 ppm
- No Class 1,2,3 or other solvents are used or produced in the:= 5 ppm
Ergebnisse des Spezifikationstests
Über diesen Artikel
:
:294.1 g/mol
Merck-Index:13,08675
Schmelzender Platingehalt:150 °C
Dichte:1.76 g/cm³ (20 °C)
Lagertemperatur:Ambient
EINECS:200-675-3
CAS-Nummer:6132-04-3
Formel:Na₃C₆H₅O₇·2H₂O
MDL-Nummer:MFCD00150031