
Citric acid monohydrate ≥99.9998% (metals basis), TraceSELECT™, Fluka™
Catalog # 94068-100G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Nitrate (NO₃):Max. 10000 ppb
- Phosphate (PO₄):Max. 2 ppm
- Sulfate (SO₄):Max. 10000 ppb
- Acetate (C₂H₃O₂):Max. 5000 ppb
- Formate (CHO₂):Max. 5000 ppb
- Glycolate (C₂H₃O₃):Max. 5000 ppb
- Oxalate (C₂O₄):Max. 5000 ppb
- Total N:Max. 5 ppm
- Tartrate:Max. 0.2 %
- Assay:99.5 - 101.0 %
- Silver (Ag):Max. 0.01 ppm
- Aluminium (Al):Max. 0.05 ppm
- Arsenic (As):Max. 0.01 ppm
- Gold (Au):Max. 0.01 ppm
- Barium (Ba):Max. 0.05 ppm
- Beryllium (Be):Max. 0.01 ppm
- Bismuth (Bi):Max. 0.01 ppm
- Calcium (Ca):Max. 0.1 ppm
- Cadmium (Cd):Max. 0.01 ppm
- Cerium (Ce):Max. 0.01 ppm
- Cobalt (Co):Max. 0.01 ppm
- Chromium (Cr):Max. 0.05 ppm
- Cesium (Cs):Max. 0.01 ppm
- Copper (Cu):Max. 0.01 ppm
- Dysprosium (Dy):Max. 0.01 ppm
- Erbium (Er):Max. 0.01 ppm
- Europium (Eu):Max. 0.01 ppm
- Iron (Fe):Max. 0.1 ppm
- Gadolinium (Gd):Max. 0.01 ppm
- Germanium (Ge):Max. 0.01 ppm
- Hafnium (Hf):Max. 0.01 ppm
- Mercury (Hg):Max. 5 ppb
- Holmium (Ho):Max. 0.01 ppm
- Indium (In):Max. 0.01 ppm
- Iridium (Ir):Max. 0.01 ppm
- Potassium (K):Max. 0.1 ppm
- Lanthanum (La):Max. 0.01 ppm
- Lithium (Li):Max. 0.01 ppm
- Lutetium (Lu):Max. 0.01 ppm
- Magnesium (Mg):Max. 0.05 ppm
- Manganese (Mn):Max. 0.01 ppm
- Molybdenum (Mo):Max. 0.01 ppm
- Sodium (Na):Max. 2 ppm
- Niobium (nb):Max. 0.01 ppm
- Neodymium (nd):Max. 0.01 ppm
- Nickel (Ni):Max. 0.01 ppm
- Lead (Pb):Max. 0.05 ppm
- Palladium (Pd):Max. 0.01 ppm
- Praseodymium (Pr):Max. 0.01 ppm
- Platinum (Pt):Max. 0.01 ppm
- Rubidium (Rb):Max. 0.01 ppm
- Rhodium (Rh):Max. 0.01 ppm
- Ruthenium (Ru):Max. 0.01 ppm
- Antimony (Sb):Max. 0.01 ppm
- Scandium (Sc):Max. 0.05 ppm
- Selenium (Se):Max. 0.05 ppm
- Samarium (Sm):Max. 0.01 ppm
- Tin (Sn):Max. 0.01 ppm
- Strontium (Sr):Max. 0.01 ppm
- Tantalum (Ta):Max. 0.01 ppm
- Terbium (Tb):Max. 0.01 ppm
- Tellurium (Te):Max. 0.01 ppm
- Thorium (Th):Max. 0.01 ppm
- Titanium (Ti):Max. 0.05 ppm
- Thallium (Tl):Max. 0.01 ppm
- Thulium (Tm):Max. 0.01 ppm
- Uran (U):Max. 0.01 ppm
- Vanadium (V):Max. 0.01 ppm
- Tungsten (W):Max. 0.01 ppm
- Yttrium (Y):Max. 0.01 ppm
- Ytterbium (Yb):Max. 0.01 ppm
- Zinc (Zn):Max. 0.01 ppm
- Zirconium (Zr):Max. 0.01 ppm
- Bromide (Br):Max. 2000 ppb
- Chloride (Cl):Max. 10000 ppb
- Fluoride (F):Max. 2000 ppb
- Iodide (I):Max. 2000 ppb
- Nitrite (NO₂):Max. 2000 ppb
Specification Test Results
About this item
:
:210.14 g/mol
Merck Index:13,02350
Melting Pt:135-152 °C
Density:1.54 g/cm³ (20 °C)
Flash Pt:173,9 °C (closed cup)
EINECS:201-069-1
:GHS07
Storage Temperature:Ambient
CAS Number:5949-29-1
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972
Melting Pt:135…152 °C