Order Entry
Switzerland
ContactUsLinkComponent
 
 
Oxalic acid dihydrate ≥99%, GPR RECTAPUR®
 
undefined
 
Oxalic acid dihydrate ≥99%, GPR RECTAPUR®
 

 

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 0.05 %
  • Fe (Iron):
    Max. 20 ppm

 

Frequently Bought Together