

Oxalic acid dihydrate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6153-56-6
:
Formula:HO₂CCO₂H·2H₂O
MDL Number:MFCD00149102
:126.07 g/mol
Merck Index:13,06980
Boiling Pt:149…160 °C (1013 hPa)
Melting Pt:101 °C
Density:1.65 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3261
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:205-634-3
REACH:01-2119534576-33
:GHS05
:GHS07
Specification Test Results
- Solubility:102 g/l (20 °C)
- Melting Point:101 °C
- Molar Mass:126.07 g/mol
- Bulk Density:900 kg/m3
- Boiling Point:149 - 160 °C (decomposition)
- Vapor Pressure:21 hPa (50 °C)
- Flash Point:157 °C (decomposition)
- Density:1.65 g/cm3 (20 °C)
- pH:1 (10 g/l, H2O, 20 °C)
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.18242.1000_TDS_EN ProductPage_818242
Product specification
msds_818242
SDS
Frequently Bought Together