Zinc acetate dihydrate, EMSURE® ACS for analysis, Supelco®
: Merck
:5970-45-6
:
:(H₃CCOO)₂Zn·2H₂O
:MFCD00066961
:219.51 g/mol
:12,10256
:237 °C
:1.718 g/cm³ (20 °C)
:Ambient
:209-170-2
:9,III
:M-Clarity™ quality level = MQ300
:3077
:GHS05
:GHS07
:GHS09
- Solubility:430 g/l (20 °C)
- Melting Point:237 °C
- Molar Mass:219.49 g/mol
- Bulk Density:900 kg/m3
- Density:1.74 g/cm3 (20 °C)
- pH:6 - 7 (50 g/l, H2O, 20 °C)
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Frequently Bought Together