Order Entry
Switzerland
ContactUsLinkComponent
Cadmium acetate dihydrate, EMSURE® for analysis, Supelco®
Cadmium acetate dihydrate, EMSURE® for analysis, Supelco®
Catalog # 1.02003.0500
Supplier:  Merck
CAS Number:  
Cadmium acetate dihydrate, EMSURE® for analysis, Supelco®
Catalog # 1.02003.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    256 °C (anhydrous)
  • Molar Mass:
    266.52 g/mol
  • Bulk Density:
    950 kg/m3
  • Density:
    2.01 g/cm3 (20 °C)
  • pH:
    7 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

Specification Test Results

About this item

Synonyms: Cadmium di(acetate) dihydrate, Acetic acid cadmium salt dihydrate
 :
 :266.53 g/mol
Merck Index:13,01614
Boiling Pt:105…107 °C (4 mmHg)
Melting Pt:256 °C
Density:2.341 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-853-2
UN:2570
ADR:6.1,II
 :M-Clarity™ quality level = MQ300
 :GHS06
 :GHS08
 :GHS09
CAS Number:5743-04-4
Formula:(H₃CCOO)₂Cd·2H₂O
MDL Number:MFCD00150020