



Hydroxylammonium chloride ≥96% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5470-11-1
:
Formula:NH₂OH·HCl
MDL Number:MFCD00051089
:69.49 g/mol
Merck Index:14,4828
Boiling Pt:306 °C (1013 hPa)
Melting Pt:152 °C (dec.)
Density:1.67
EINECS:226-798-2
UN:2923
ADR:8,III
:GHS08
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together