



Hydroxylammonium chloride 97%
:5470-11-1
:
:NH₂OH·HCl
:MFCD00051089
:69.49 g/mol
:13,04853
:306 °C (1013 hPa)
:155…158 °C (dec.)
:1,67 g/cm³ (20 °C)
:226-798-2
:2923
:8,III
:GHS05
:GHS06
:GHS08
:GHS09
:Ambient
- Appearance:White to light yellow Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:151 to 157 °C (Dec)
- Titration with NaOH:≥96.0 %
Frequently Bought Together