



Hydroxylammonium sulphate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10039-54-0
:
Formula:H₈N₂O₆S
MDL Number:MFCD00044869
:164.14 g/mol
Merck Index:14,4828
Melting Pt:∼ 170 °C (dec.)
Density:1.88
EINECS:233-118-8
UN:2865
ADR:8,III
:GHS08
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together