ContactUsLinkComponent
IHC enhancer (DAB, plus), HIGHDEF®
IHC enhancer (DAB, plus), HIGHDEF®
Catalog # ENZOADI9501820015
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
IHC enhancer (DAB, plus), HIGHDEF®
Catalog # ENZOADI9501820015
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Pk:
    15 mL
  • Description:
    HIGHDEF® IHC enhancer (DAB, plus), 1X
  • Seq:
    02
Specifications
About this item
HIGHDEF® IHC enhancer (DAB, plus) is a DAB signal enhancer used to increase intensity of immunostaining using DAB as chromogen.
  • Superior performance compared to amino ethyl carbazol (AEC)
HIGHDEF® IHC enhancer (DAB, plus) increases the signal intensity without creating any background staining.