


4-(Methylamino)phenol sulphate 99.0-101.5% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:55-55-0
:51-72-9
Formula:C₁₄H₂₀N₂O₆S
MDL Number:MFCD00013140
:344.39 g/mol
Merck Index:14,6017
Boiling Pt:227 °C (1013 hPa)
Melting Pt:∼. 260 °C (dec.)
Density:1.434 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-237-1
UN:2811
ADR:6.1,III
:GHS08
:GHS07
:GHS09
Related Information
Frequently Bought Together