

Silver nitrate concentrate for dilution to 1L, 0.5 M AgNO3 (0.5N) volumetric solution, Fluka™
Catalog # 38311-1EA
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Ampoule
- Pk:1
- Seq:0001
Specifications
Specification Test Results
- Factor:0.998 - 1.002
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Specification Test Results
About this item
:
:169.87 g/mol
Boiling Pt:100 °C
Storage Temperature:Ambient
EINECS:231-853-9
UN:3264
ADR:8,II
:GHS05
:GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414