Order Entry
Switzerland
ContactUsLinkComponent
 
Ammonium thiocyanate concentrate for dilution to 1L, 0.1 M NH4SCN (0.1N) volumetric solution, Fluka™
  38020-1EA
 :  
Ammonium thiocyanate concentrate for dilution to 1L, 0.1 M NH4SCN (0.1N) volumetric solution, Fluka™
  38020-1EA
 :  

 

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

 

 

  • Factor:
    0.998 - 1.002

 

 

 :
 :76.12 g/mol
 :100 °C (1.013 hPa)
 :Ambient
 :217-175-6
 :1762-95-4
 :NH₄SCN
 :MFCD00011428