Order Entry
Switzerland
ContactUsLinkComponent
 
Corrosive
Potassium iodate 1/60 mol/l (3.567 g KIO₃) concentrated aqueous solution, Titrisol® volumetric solution, Supelco®
Catalog # 1.09917.0001
Supplier:  Merck
CAS Number:  
Corrosive
Potassium iodate 1/60 mol/l (3.567 g KIO₃) concentrated aqueous solution, Titrisol® volumetric solution, Supelco®
Catalog # 1.09917.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Density:
    1.03 g/cm3 (20 °C)
  • pH:
    5.4 (H2O, 20 °C)

Specification Test Results

About this item

 :
 :214 g/mol
ADR:5.1,II
 :M-Clarity™ quality level = MQ100
EINECS:231-831-9
UN:3139
 :GHS05
Storage Temperature:Ambient
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406