Order Entry
Switzerland
ContactUsLinkComponent
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Catalog # EURLEMR256005BK
Supplier:  EUROCLONE
CAS Number:  
Buffer, TBE solution 5X concentrate (TRIS-borate-EDTA buffer)
Catalog # EURLEMR256005BK
Supplier:  EUROCLONE
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer)

Specifications

About this item