Buffer, TBE solid for 10X concentrated solution (TRIS-borate-EDTA buffer)
TBE buffer solid for 10X concentrated solution (TRIS-borate-EDTA buffer), 1 KIT
Catalog # EURLEMR042004
Supplier: EUROCLONE
Buffer, TBE solid for 10X concentrated solution (TRIS-borate-EDTA buffer)
Catalog # EURLEMR042004
Supplier: EUROCLONE
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:1 KIT
- Description:TBE buffer solid for 10X concentrated solution (TRIS-borate-EDTA buffer)
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



