Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer), Glass bottle, 1 L
Catalog # NADIEC-872-1L
Marketsource
Supplier: NATIONAL DIAGNOSTICS
Buffer, TAE solution 50X concentrate (TRIS-acetate-EDTA buffer)
Catalog # NADIEC-872-1L
Supplier: NATIONAL DIAGNOSTICS
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Description:TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)



