Ethidium bromide 10 mg/ml in aqueous solution
Glass bottle, 25 mL
Catalog # EURLEMR035025
Supplier: EUROCLONE
Ethidium bromide 10 mg/ml in aqueous solution
Catalog # EURLEMR035025
Supplier: EUROCLONE
CAS Number: 1239-45-8
Jump to:
Product Details & Documents
Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
- CAS Number:1239-45-8
- Formula:C₂₁H₂₀BrN₃
- MDL Number:MFCD00011724
- Molecular Weight:394.31 g/mol
- EINECS:214-984-6
Specifications
Specifications
- Pack type:Glass bottle
- Pk:25 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



