Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Silver nitrate 10% in aqueous solution

Silver nitrate 10% in aqueous solution

Glass bottle, 500 mL
Catalog # CTEH64129/5
Corrosive
Environment
Marketsource
Supplier:   CLIN TECH
Silver nitrate 10% in aqueous solution
Catalog # CTEH64129/5
Supplier:  CLIN TECH
Marketsource
CAS Number:  7761-88-8
Silver nitrate 10% in aqueous solution
Silver nitrate 10% in aqueous solution

Glass bottle, 500 mL

CAS Number:  7761-88-8
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • CAS Number:7761-88-8
  • Formula:AgNO₃
  • MDL Number:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-853-9
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications