



Hydrazine hydrate 55% in aqueous solution (∼35% w/w N₂H₄)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10217-52-4
:7803-57-8
Formula:NH₂NH₂·xH₂O
MDL Number:MFCD00149931
:32.05 g/mol (anhy)
Melting Pt:–65 °C
Density:1.023 g/cm³
Flash Pt:>100 °C
EINECS:206-114-9
UN:3293
ADR:6.1,III
:GHS06
:GHS08
:GHS05
:GHS09
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless Solution
- Refractive index:1.3860 to 1.3900 (20°C, 589 nm)
- Density:(20°C) 1.021 to 1.025 g/ml
- Titration with HCl:35.0 to 35.6 % (N2H4)
Specification Test Results
Frequently Bought Together