Order Entry
Switzerland
ContactUsLinkComponent
Ammonium sulphide 20% in aqueous solution, GPR RECTAPUR®
Flammable
Corrosive
Ammonium sulphide 20% in aqueous solution, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Corrosive
Ammonium sulphide 20% in aqueous solution, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (as (NH4)2S from sulphide):
    17 - 23 %
  • Ignition residue (SO4):
    Max. 0.05 %

Specification Test Results

Frequently Bought Together