Ammonia solution 33%, BAKER, Laboratory Reagent, J.T.Baker®
: Avantor
:1336-21-6
:7664-41-7
:NH₃
:MFCD00066650
:35.05 g/mol
:32…37.7 °C (1013 hPa)
:–60…–57.5 °C
:0.88…0.91 g/cm³ (20 °C)
:215-647-6
:2672
:8,III
:GHS05
:GHS07
:GHS09
:Ambient
- Assay (as NH3) (acidimetric):30 - 33 %
- Chloride (Cl):max. 0.002 %
- Residue after Ignition:max. 0.003 %
- Substances Reducing KMnO4:max. 0.0008 %
- Sulfate (SO4):max. 0.003 %
- Trace Impurities (in ppm) Iron (Fe):max. 5
- Heavy Metals (as Pb):max. 1
Frequently Bought Together