SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Ammonia solution
Ammonia solution 28.0 - 30.0% ACS
Ammonia solution 28.0 - 30.0% ACS
:
Thermo Fisher Scientific
Ammonia solution 28.0 - 30.0% ACS
:
Thermo Fisher Scientific
:
1336-21-6
:
7664-41-7
:
NH₃
:
MFCD00066650
:
35.05 g/mol
:
14,494
:
32…37.7 °C (1013 hPa)
:
–60…–57.5 °C
:
0.90
:
Ambient
:
215-647-6
:
2672
:
8,III
:
GHS05
:
GHS07
:
GHS09
msds_ALFA033285
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Ethanol absolute ≥99.8%, Electran for molecular biology
: VWR Chemicals
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck
Test Strips, nitrate, MQuant®, Supelco®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
Test kits, nitrite, Spectroquant®, Supelco®
: Merck
Test strips, peroxide, Dosatest
: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
: Merck
Test strips for peroxide, MQuant®, Supelco®
: Merck
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
: VWR Chemicals
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
VWR® Rota®, pH indicator paper, 1 to 14
: VWR Chemicals
SPECTRUM REUSABLE DESICCANT PACK 1 * 1 items
: PerkinElmer