Order Entry
Switzerland
ContactUsLinkComponent
Ammonia solution 25%, GPR RECTAPUR®
Corrosive
Irritant
Environment
Ammonia solution 25%, GPR RECTAPUR®
Catalog # 271402T
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Environment
Ammonia solution 25%, GPR RECTAPUR®
Catalog # 271402T
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay:
    Min. 25 %
  • Non-volatile residue:
    Max. 100 ppm
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Pb (Lead):
    Max. 5 ppm

Specification Test Results

About this item

 :
 :35.05 g/mol
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.907 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-647-6
UN:2672
ADR:8,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650