Tetramethylammonium hydroxide 25% in aqueous solution
Catalog # WAKO206-15035
Supplier: Wako
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:91.15 g/mol
Boiling Pt:110 °C (1013 hPa)
Density:1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-882-9
UN:1835
ADR:8,II
:GHS06
:GHS05
CAS Number:75-59-2
Formula:C₄H₁₃NO
MDL Number:MFCD00008280