Order Entry
Switzerland
ContactUsLinkComponent
Tetramethylammonium hydroxide 25% in aqueous solution, GPR RECTAPUR®
Toxic
Corrosive
Tetramethylammonium hydroxide 25% in aqueous solution, GPR RECTAPUR®
Catalog # 191773B
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Corrosive
Tetramethylammonium hydroxide 25% in aqueous solution, GPR RECTAPUR®
Catalog # 191773B
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Assay:
    24 - 26 %
  • Halide (as Cl):
    Max. 0.2 %
  • Ignition residue (SO4):
    Max. 0.1 %

Specification Test Results

About this item

 :
 :91.15 g/mol
Boiling Pt:110 °C (1013 hPa)
Density:1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-882-9
UN:1835
ADR:8,II
 :GHS06
 :GHS05
CAS Number:75-59-2
Formula:C₄H₁₃NO
MDL Number:MFCD00008280