Tetramethylammonium hydroxide 25% in water
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-59-2
:
Formula:C₄H₁₃NO
MDL Number:MFCD00008280
:91.15 g/mol
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–25 °C
Density:1.014 g/cm³
Flash Pt:>95 °C
EINECS:200-882-9
UN:1835
ADR:8,II
:GHS06
:GHS05
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Solution
- Refractive index:1.3780 to 1.3840 (20°C, 589 nm)
- Titration with HCl:23 to 27 %
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together