Tetraethylammonium hydroxide 35% (w/w) in aqueous solution, Reagent Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-98-5
:
Formula:C₈H₂₁NO
MDL Number:MFCD00009024
:147.26 g/mol
Merck Index:14,9201
Boiling Pt: Dec.
Density:1.023
Storage Temperature:Ambient
EINECS:201-073-3
UN:3267
ADR:8,II
:GHS05
Frequently Bought Together