



Tetraethylammonium hydroxide 25% (w/w) in methanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:77-98-5
:
Formula:C₈H₂₁NO
MDL Number:MFCD00009024
:147.26 g/mol
Merck Index:14,9201
Boiling Pt: Dec.
Density:0.88
Flash Pt:16 °C (60 °F)
EINECS:201-073-3
UN:2924
ADR:3,II
:GHS02
:GHS06
:GHS08
:GHS05
Storage Temperature:Ambient
Frequently Bought Together