Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
Tetraethylammonium hydroxide 20% (w/w) in water
 
undefined
 
 
 
Tetraethylammonium hydroxide 20% (w/w) in water

 

  • Appearance:
    Clear colorless to light yellow Solution
  • Titration with HCl:
    ≥20 %
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together