Order Entry
Switzerland
ContactUsLinkComponent
Tetraethylammonium hydroxide 10% in aqueous solution
Corrosive
Toxic
Health
Tetraethylammonium hydroxide 10% in aqueous solution
Certificates
undefined
Corrosive
Toxic
Health
Tetraethylammonium hydroxide 10% in aqueous solution

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • Titration with HCl:
    9.5 to 10.5 %
  • Melting point:
    153 to 157 °C
  • Titration with NaOH:
    ≥98.0 %

Specification Test Results

Related Information

Frequently Bought Together