ContactUsLinkComponent
Silver sulphate 6,6 g/l in sulphuric acid, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Corrosive
Silver sulphate 6,6 g/l in sulphuric acid, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Silver sulphate 6,6 g/l in sulphuric acid, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Chemical oxygen demand (COD):
    Passes test
  • NIST traceable:
    Confirmed