Silver sulphate 6.6 g/l in sulphuric acid, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 7641.2500
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:311.8 g/mol
EINECS:233-653-7
UN:1830
ADR:8,II
:GHS05
CAS Number:10294-26-5
Formula:Ag₂SO₄
MDL Number:MFCD00003407