
Ammonium cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 2678.1000
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.0998 - 0.1002 mol/l
- NIST traceable:Confirmed
Specification Test Results
About this item
:
:596.52
EINECS:231-567-4
UN:3264
ADR:8,III
:GHS05
CAS Number:7637-03-8
Formula:H₁₆CeN₄O₁₆S₄
MDL Number:MFCD00148851