ContactUsLinkComponent
Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur., Reag. USP volumetric solution
Corrosive
Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur., Reag. USP volumetric solution
Supplier:  VWR Chemicals
Certificates
undefined
Corrosive
Cerium(IV) sulphate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur., Reag. USP volumetric solution
Supplier:  VWR Chemicals
Certificates
Specification Test Results
  • Titer (20°C; real value 0.3 % accuracy):
    0,0995-0,1005 mol/l
  • NIST traceable:
    Confirmed
  • Conforms to Reag. Ph.Eur.:
    Passes test
  • Conforms to Reag. USP:
    Passes test