Order Entry
Switzerland
ContactUsLinkComponent
Cerium(IV) sulphate 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Irritant
Cerium(IV) sulphate 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31307.294
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Cerium(IV) sulphate 0.05 mol/l (0.05 N) in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 31307.294
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    0.0499 - 0.0501 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :332.24 g/mol
Merck Index:13,02002
EINECS:237-029-5
UN:3264
ADR:8,III
 :GHS07
CAS Number:13590-82-4
Formula:Ce(SO₄)₂
MDL Number:MFCD00148852