



Tetrabutylammonium hydroxide 1.0 M in methanol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2052-49-5
:
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425
:259.48 g/mol
Density:0.830
Flash Pt:11 °C (51 °F)
Storage Temperature:Ambient
EINECS:218-147-6
UN:2924
ADR:3,II
:GHS02
:GHS06
:GHS08
:GHS05
Related Information
Frequently Bought Together