ContactUsLinkComponent
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Corrosive
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Certificates
undefined
Corrosive
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Specification Test Results
  • Appearance:
    Clear solution
  • UV:
    1 cm cell vs water
  • Color scale:
    ≤10 APHA
  • Titration with HCl:
    0.37 to 0.43 M
  • Bromide (Br):
    ≤500 ppm