Order Entry
Switzerland
ContactUsLinkComponent
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in toluene/methanol for titration in non-aqueous media
Flammable
Irritant
Health
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in toluene/methanol for titration in non-aqueous media
Certificates
undefined
Flammable
Irritant
Health
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in toluene/methanol for titration in non-aqueous media

Specification Test Results

  • Appearance:
    Clear colorless to yellow Solution
  • Titration with HClO4:
    0.09 to 0.11 M
  • Melting point:
    52 to 56 °C
  • GC:
    ≥98.5 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

Related Information

Frequently Bought Together