Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
Hydrochloric acid 37%, SPECTRONORM® for atomic absorption spectroscopy
  141577U
 :  
undefined
 
 
Hydrochloric acid 37%, SPECTRONORM® for atomic absorption spectroscopy
  141577U
 :  

 

  • Pack type:
    Glass bottle, Safebreak®
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Al (Aluminium):
    Max. 0.05 ppm
  • Ba (Barium):
    Max. 0.02 ppm
  • Ca (Calcium):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.01 ppm
  • Co (Cobalt):
    Max. 0.01 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.02 ppm
  • Fe (Iron):
    Max. 0.2 ppm
  • Hg (Mercury):
    Max. 0.005 ppm
  • K (Potassium):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.01 ppm
  • Na (Sodium):
    Max. 0.5 ppm
  • Ni (Nickel):
    Max. 0.02 ppm
  • Pb (Lead):
    Max. 0.02 ppm
  • Sr (Strontium):
    Max. 0.01 ppm
  • Zn (Zinc):
    Max. 0.05 ppm

 

 

 :
 :110 °C (1013 hPa)
 :–30 °C
 :1.18 g/cm³ (20 °C)
 :1789
 :8,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
 :36.46 g/mol
 :Ambient
 :231-595-7
 :01-2119484862-27
 :GHS05
 :GHS07
 :7647-01-0
 :HCl
 :MFCD00011324