Order Entry
Switzerland
ContactUsLinkComponent
Sodium hydroxide 0.2 M NaOH (0.2N) volumetric solution, Fluka™
Corrosive
Sodium hydroxide 0.2 M NaOH (0.2N) volumetric solution, Fluka™
Certificates
Corrosive
Sodium hydroxide 0.2 M NaOH (0.2N) volumetric solution, Fluka™

Specification Test Results

  • Assay:
    0.1900 - 0.2100 N
  • Carbonate (as Na₂CO₃):
    Max. 0.006 N
  • Traceable to NIST SRM:
    effective

Specification Test Results

Related Information

Frequently Bought Together